Anti-CA8 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50767-T60-50 50767-T60-200 50767-T60-100
103589-302EA 106.62 USD
103589-302 103595-654 103595-652
Anti-CA8 Rabbit Polyclonal Antibody
Antibodies
Anti-CA8 Antibody

Type: Primary
Antigen: CA8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR