Anti-CA8 Rabbit Monoclonal Antibody [clone:057]

Supplier: Sino Biological
50767-R057-1 50767-R057-100 50767-R057-50
103658-940EA 271.4 USD
103658-940 103589-300 103595-650
Anti-CA8 Rabbit Monoclonal Antibody [clone:057]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase VIII / CA8 (rM Carbonic Anhydrase VIII / CA8; Catalog#50767-M08E; P28651; Met1-Gln291).

Type: Primary
Antigen: CA8
Clonality: Monoclonal
Clone: 057
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR