Anti-CA8 Rabbit Monoclonal Antibody [clone:005]

Supplier: Sino Biological
50767-R005-50 50767-R005-100
103595-648EA 150.18 USD
103595-648 103658-938
Anti-CA8 Rabbit Monoclonal Antibody [clone:005]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase VIII / CA8 (rM Carbonic Anhydrase VIII / CA8; Catalog#50767-M08E; P28651; Met1-Gln291).

Type: Primary
Antigen: CA8
Clonality: Monoclonal
Clone: 005
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR