Anti-LILRB3 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50760-T16-50 50760-T16-200 50760-T16-100
103589-296EA 106.62 USD
103589-296 103595-644 103595-642
Anti-LILRB3 Rabbit Polyclonal Antibody
Antibodies
Anti-LILRB3 Antibody

Type: Primary
Antigen: LILRB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR