Anti-LILRB3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50760-RP01-200 50760-RP01-100 50760-RP01-400
103658-930EA 351.49 USD
103658-930 103658-934 103658-932
Anti-LILRB3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LILRB3 (Catalog#50760-M08H; AAC53219.1; Met1-Tyr640). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LILRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR