Anti-CD79B Rabbit Polyclonal Antibody

Supplier: Sino Biological

50754-T16-100 50754-T16-50 50754-T16-200
103595-618EA 160.46 USD
103595-618 103589-286 103595-620
Anti-CD79B Rabbit Polyclonal Antibody
Antibodies
Anti-CD79B Antibody

Type: Primary
Antigen: CD79B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR