Anti-LTF Rabbit Polyclonal Antibody

Supplier: Sino Biological

50750-RP01-400 50750-RP01-100 50750-RP01-200
103659-508EA 214.31 USD
103659-508 103659-506 103659-504
Anti-LTF Rabbit Polyclonal Antibody
Antibodies
Anti-Lactoferrin Antibody

Type: Primary
Antigen: LTF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR