Anti-SELP Rabbit Polyclonal Antibody

Supplier: Sino Biological
50737-T16-200 50737-T16-100 50737-T16-50
103595-606EA 352.03 USD
103595-606 103595-604 103589-280
Anti-SELP Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse SELP / P Selectin (rM SELP / P Selectin; Catalog#50737-M08H; Q01102; Met1-Ala709). SELP / P Selectin specific IgG was purified by Mouse SELP / P Selectin affinity chromatography.

Type: Primary
Antigen: SELP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR