Anti-SELP Rabbit Polyclonal Antibody

Supplier: Sino Biological

50737-RP01-100 50737-RP01-400 50737-RP01-200
103659-496EA 106.62 USD
103659-496 103659-500 103659-498
Anti-SELP Rabbit Polyclonal Antibody
Antibodies
Anti-P-Selectin Antibody

Type: Primary
Antigen: SELP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR