Anti-CD69 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50731-T16-50 50731-T16-200 50731-T16-100
103589-276EA 106.62 USD
103589-276 103595-596 103595-594
Anti-CD69 Rabbit Polyclonal Antibody
Antibodies
Anti-CD69 Antibody

Type: Primary
Antigen: CD69
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR