Anti-SMAD5 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50728-RP01-200 50728-RP01-100 50728-RP01-400
103658-902EA 160.46 USD
103658-902 103658-904 103658-906
Anti-SMAD5 Rabbit Polyclonal Antibody
Antibodies
Anti-Smad5 Antibody

Type: Primary
Antigen: SMAD5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR