Anti-SDC4 Rabbit Monoclonal Antibody [clone: 047]

Supplier: Sino Biological

50726-R047-100 50726-R047-50
103658-898EA 214.31 USD
103658-898 103595-580
Anti-SDC4 Rabbit Monoclonal Antibody [clone: 047]
Antibodies
Anti-Syndecan-4 Antibody

Type: Primary
Antigen: SDC4
Clonality: Monoclonal
Clone: Clone: 047
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR