Anti-SDC4 Rabbit Monoclonal Antibody [clone: 008]

Supplier: Sino Biological

50726-R008-100 50726-R008-50
103658-896EA 214.31 USD
103658-896 103595-578
Anti-SDC4 Rabbit Monoclonal Antibody [clone: 008]
Antibodies
Anti-Syndecan-4 Antibody

Type: Primary
Antigen: SDC4
Clonality: Monoclonal
Clone: Clone: 008
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR