Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD59a Rabbit Polyclonal Antibody

Supplier: Sino Biological

50724-T24-100 50724-T24-50 50724-T24-200
103595-574EA 160.46 USD InStock
103595-574 103589-270 103595-576
Anti-CD59a Rabbit Polyclonal Antibody
Antibodies
Anti-CD59a Antibody

Type: Primary
Antigen: CD59a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty