Anti-CD59a Rabbit Polyclonal Antibody

Supplier: Sino Biological
50724-RP01-100 50724-RP01-200 50724-RP01-400
103658-894EA 562.97 USD
103658-894 103658-892 103658-890
Anti-CD59a Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD59a / Protectin (Catalog#50724-M08H; NP_031680.2; Met1-Lys95). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD59a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR