Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD59a Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 133]

Supplier: Sino Biological

50724-R133-F-100 50724-R133-F-25
103632-330EA 258.46 USD InStock
103632-330 103631-438
Anti-CD59a Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 133]
Antibodies
Anti-CD59a Antibody (FITC)

Type: Primary
Antigen: CD59a
Clonality: Monoclonal
Clone: Clone: 133
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty