Anti-CD59a Rabbit Monoclonal Antibody [clone:133] APC (Allophycocyanin)

Supplier: Sino Biological
50724-R133-A-25 50724-R133-A-100
103632-328EA 282.8 USD
103632-328 103631-436
Anti-CD59a Rabbit Monoclonal Antibody [clone:133] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD59a / Protectin (rM CD59a / Protectin; Catalog#50724-M08H; NP_031680.2; Met1-Lys95) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD59a
Clonality: Monoclonal
Clone: Clone: 133
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR