Anti-CD59a Rabbit Monoclonal Antibody [clone:045]

Supplier: Sino Biological
50724-R045-100 50724-R045-50
103595-570EA 150.18 USD
103595-570 103658-886
Anti-CD59a Rabbit Monoclonal Antibody [clone:045]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD59a / Protectin (rM CD59a / Protectin; Catalog#50724-M08H; NP_031680.2; Met1-Lys95).

Type: Primary
Antigen: CD59a
Clonality: Monoclonal
Clone: 045
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR