Anti-CD53 Rabbit Monoclonal Antibody [clone:013]

Supplier: Sino Biological
50722-R013-50 50722-R013-100
103658-878EA 271.4 USD
103658-878 103595-564
Anti-CD53 Rabbit Monoclonal Antibody [clone:013]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD53 (rM CD53; Catalog#50722-M07H; Q61451; Glu107-Asn181).

Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: 013
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR