Anti-Her2 / ERBB2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50714-RP01-100 50714-RP01-400 50714-RP01-200
103658-866EA 106.62 USD
103658-866 103658-870 103658-868
Anti-Her2 / ERBB2 Rabbit Polyclonal Antibody
Antibodies
Anti-Her2 / ERBB2 Antibody

Type: Primary
Antigen: Her2 / ERBB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR