Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Anti-CD33 Antibody Rabbit Polyclonal Antibody

Supplier: Sino Biological

50712-T16-200 50712-T16-100 50712-T16-50
103595-544EA 214.31 USD InStock
103595-544 103595-542 103589-260
Anti-Anti-CD33 Antibody Rabbit Polyclonal Antibody
Antibodies
Anti-CD33 Antibody

Type: Primary
Antigen: Anti-CD33 Antibody
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty