Anti-CD6 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50711-RP02-100 50711-RP02-200 50711-RP02-50
103659-490EA 160.46 USD
103659-490 103659-492 103595-540
Anti-CD6 Rabbit Polyclonal Antibody
Antibodies
Anti-CD6 Antibody

Type: Primary
Antigen: CD6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR