Anti-CD6 Rabbit Monoclonal Antibody [clone:309]

Supplier: Sino Biological
50711-R309-100 50711-R309-50
103658-850EA 271.4 USD
103658-850 103595-538
Anti-CD6 Rabbit Monoclonal Antibody [clone:309]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD6 (rM CD6; Catalog#50711-M08H; Q91WN5; Met1-Gly396).

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 309
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR