Anti-CD6 Rabbit Monoclonal Antibody [clone: 039]

Supplier: Sino Biological
50711-R039-1 50711-R039-100 50711-R039-50
103595-536EA 119 USD
103595-536 103589-258 103658-848
Anti-CD6 Rabbit Monoclonal Antibody [clone: 039]
Antibodies
Anti-CD6 Antibody

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: Clone: 039
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR