Anti-CD6 Rabbit Monoclonal Antibody [clone:002]

Supplier: Sino Biological
50711-R002-1 50711-R002-50 50711-R002-100
103589-256EA 1500.59 USD
103589-256 103595-532 103658-844
Anti-CD6 Rabbit Monoclonal Antibody [clone:002]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD6 / Cluster of Differentiation 6 (rM CD6 / Cluster of Differentiation 6; Catalog#50711-M08H; Q91WN5; Met1-Gly396).

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 002
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR