Anti-CLEC10A Rabbit Polyclonal Antibody

Supplier: Sino Biological
50707-T16-100 50707-T16-200 50707-T16-50
103595-518EA 352.03 USD
103595-518 103589-250 103595-516
Anti-CLEC10A Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CLEC10A / MGL1 / CD301 (Catalog#50707-M07H; NP_001191181.1; Gln58-Ser305). CLEC10A / MGL1 / CD301 specific IgG was purified by Mouse CLEC10A / MGL1 / CD301 affinity chromatography.

Type: Primary
Antigen: CLEC10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR