Anti-CLEC10A Rabbit Polyclonal Antibody

Supplier: Sino Biological
50707-RP01-200 50707-RP01-400 50707-RP01-100
103660-718EA 351.49 USD
103660-718 103660-716 103660-720
Anti-CLEC10A Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CLEC10A / MGL1 / CD301 (Catalog#50707-M07H; NP_001191181.1; Gln58-Ser305). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CLEC10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR