Anti-CLEC10A Rabbit Monoclonal Antibody [clone:019]

Supplier: Sino Biological
50707-R019-50 50707-R019-100
103595-514EA 163.15 USD
103595-514 103660-714
Anti-CLEC10A Rabbit Monoclonal Antibody [clone:019]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CLEC10A / MGL1 / CD301 (rM CLEC10A / MGL1 / CD301; Catalog#50707-M07H; NP_001191181.1; Gln58-Ser305).

Type: Primary
Antigen: CLEC10A
Clonality: Monoclonal
Clone: 019
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR