Anti-IFNGR1 Rabbit Monoclonal Antibody [clone:062]

Supplier: Sino Biological
50705-R062-1 50705-R062-100 50705-R062-50
103589-242EA 1631.49 USD
103589-242 103658-822 103595-500
Anti-IFNGR1 Rabbit Monoclonal Antibody [clone:062]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFNGR1 / CD119 (rM IFNGR1 / CD119; Catalog#50705-M08H; P15261; Met1-Asp253).

Type: Primary
Antigen: IFNGR1
Clonality: Monoclonal
Clone: 062
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR