Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLEC3B Rabbit Polyclonal Antibody

Supplier: Sino Biological

50700-RP01-400 50700-RP01-200 50700-RP01-100
103658-816EA 214.31 USD InStock
103658-816 103658-812 103658-814
Anti-CLEC3B Rabbit Polyclonal Antibody
Antibodies
Anti-Tetranectin Antibody

Type: Primary
Antigen: CLEC3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty