Anti-CLEC3B Rabbit Monoclonal Antibody [clone: 011]

Supplier: Sino Biological

50700-R011-50 50700-R011-100
103595-496EA 128.15 USD
103595-496 103658-810
Anti-CLEC3B Rabbit Monoclonal Antibody [clone: 011]
Antibodies
Anti-Tetranectin Antibody

Type: Primary
Antigen: CLEC3B
Clonality: Monoclonal
Clone: Clone: 011
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR