Anti-CHODL Rabbit Polyclonal Antibody

Supplier: Sino Biological

50699-T24-50 50699-T24-100 50699-T24-200
103588-720EA 106.62 USD
103588-720 103595-492 103595-494
Anti-CHODL Rabbit Polyclonal Antibody
Antibodies
Anti-CHODL Antibody

Type: Primary
Antigen: CHODL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR