Anti-TGFB1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50698-T48-200 50698-T48-100 50698-T48-50
103588-718EA 142.96 USD
103588-718 103595-490 103595-488
Anti-TGFB1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse TGF-beta 1 / TGFB1 (rM TGF-beta 1 / TGFB1; Catalog#50698-M08H; P04202; Met1-Ser390). TGF-beta 1 / TGFB1 specific IgG was purified by Mouse TGF-beta 1 / TGFB1 affinity chromatography.

Type: Primary
Antigen: TGFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR