Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EMAP-II Rabbit Polyclonal Antibody

Supplier: Sino Biological

50697-RP01-100 50697-RP01-200 50697-RP01-400
103658-800EA 106.62 USD InStock
103658-800 103658-798 103658-802
Anti-EMAP-II Rabbit Polyclonal Antibody
Antibodies
Anti-EMAP-II Antibody

Type: Primary
Antigen: EMAP-II
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty