Anti-FCER2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50695-RP01-200 50695-RP01-100 50695-RP01-400
103659-476EA 270.2 USD
103659-476 103659-478 103659-480
Anti-FCER2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD23 / FCER2 (rM CD23 / FCER2; Catalog#50695-M07H; P20693-1; Glu 51-Pro 331). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR