Anti-FCER2 Rabbit Monoclonal Antibody [clone:011]

Supplier: Sino Biological
50695-R011-100 50695-R011-50
103595-480EA 150.18 USD
103595-480 103659-474
Anti-FCER2 Rabbit Monoclonal Antibody [clone:011]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD23 / FCER2 (rM CD23 / FCER2; Catalog#50695-M07H; P20693-1; Glu51-Pro331).

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: 011
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR