Anti-CD23 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 007]

Supplier: Sino Biological

50695-R007-P-100 50695-R007-P-25
103632-300EA 150.77 USD
103632-300 103630-136
Anti-CD23 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 007]
Antibodies
Anti-CD23 Antibody (PE)

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: Clone: 007
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR