Anti-CD23 Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 007]

Supplier: Sino Biological

50695-R007-C-25 50695-R007-C-100
103630-132EA 70 USD
103630-132 103632-296
Anti-CD23 Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 007]
Antibodies
Anti-CD23 Antibody (PerCP)

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: Clone: 007
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR