Anti-TCN2 Rabbit Monoclonal Antibody [clone:049]

Supplier: Sino Biological
50693-R049-100 50693-R049-50
103658-782EA 271.4 USD
103658-782 103595-470
Anti-TCN2 Rabbit Monoclonal Antibody [clone:049]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TCN2 (rM TCN2; Catalog#50693-M08H; O88968; Met1-Trp430).

Type: Primary
Antigen: TCN2
Clonality: Monoclonal
Clone: 049
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR