Anti-UCHL3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50692-RP02-100 50692-RP02-50 50692-RP02-200
103595-468EA 142.96 USD
103595-468 103658-778 103658-780
Anti-UCHL3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse UCHL3 (rM UCHL3; Catalog#50692-M07E; Q9JKB1; Glu 2-Ala 230). UCHL3 specific IgG was purified by Mouse UCHL3 affinity chromatography.

Type: Primary
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR