Anti-UCHL3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50692-RP01-200 50692-RP01-100 50692-RP01-400
103658-772EA 270.2 USD
103658-772 103658-776 103658-774
Anti-UCHL3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse UCHL3 (Catalog#50692-M07E; Q9JKB1; Glu2-Ala230). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR