Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50690-RP02-200 50690-RP02-100 50690-RP02-50
103658-762EA 270.87 USD
103658-762 103595-460 103658-764
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse UCHL1 (rM UCHL1; Catalog#50690-M07E; Q9R0P9; Gln 2-Ala 223). UCHL1 specific IgG was purified by Mouse UCHL1 affinity chromatography.

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR