Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50690-RP01-100 50690-RP01-200 50690-RP01-400
103658-756EA 106.62 USD InStock
103658-756 103658-758 103658-760
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Anti-UCHL1 Antibody

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty