Anti-CA2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50685-RP01-200 50685-RP01-100 50685-RP01-400
103658-744EA 160.46 USD
103658-744 103658-742 103658-746
Anti-CA2 Rabbit Polyclonal Antibody
Antibodies
Anti-Carbonic anhydrase II Ant

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR