Anti-AGO2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50683-RP02-100 50683-RP02-50 50683-RP02-200
103595-448EA 142.96 USD
103595-448 103658-736 103658-734
Anti-AGO2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse AGO2 (rM AGO2; Catalog#50683-M07B; Q8CJG0; Met 1-Ala 860). AGO2 specific IgG was purified by Mouse AGO2 affinity chromatography.

Type: Primary
Antigen: AGO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR