Anti-AGO2 Rabbit Monoclonal Antibody [clone: 140]

50683-R140-50 50683-R140-100
103658-732EA 214.31 USD
103658-732 103595-446
Anti-AGO2 Rabbit Monoclonal Antibody [clone: 140]
Anti-Argonaute-2 Antibody

Type: Primary
Antigen: AGO2
Clonality: Monoclonal
Clone: Clone: 140
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR