Anti-AGO2 Rabbit Monoclonal Antibody [clone:036]

Supplier: Sino Biological
50683-R036-50 50683-R036-100
103658-730EA 271.1 USD
103658-730 103595-444
Anti-AGO2 Rabbit Monoclonal Antibody [clone:036]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse AGO2 (rM AGO2; Catalog#50683-M07B; Q8CJG0; Met1-Ala860).

Type: Primary
Antigen: AGO2
Clonality: Monoclonal
Clone: 036
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR