Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Lymphotactin Rabbit Polyclonal Antibody

Supplier: Sino Biological

50677-RP01-200 50677-RP01-100 50677-RP01-400
103658-724EA 160.46 USD InStock
103658-724 103658-726 103658-728
Anti-Lymphotactin Rabbit Polyclonal Antibody
Antibodies
Anti-Lymphotactin Antibody

Type: Primary
Antigen: Lymphotactin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty