Anti-Lymphotactin Rabbit Monoclonal Antibody [clone: 002]

Supplier: Sino Biological

50677-R002-100 50677-R002-50 50677-R002-1
103660-712EA 214.31 USD
103660-712 103595-436 103589-238
Anti-Lymphotactin Rabbit Monoclonal Antibody [clone: 002]
Antibodies
Anti-Lymphotactin Antibody

Type: Primary
Antigen: Lymphotactin
Clonality: Monoclonal
Clone: Clone: 002
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR