Anti-CDH1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50671-T60-200 50671-T60-100 50671-T60-50
103595-434EA 214.31 USD
103595-434 103595-432 103589-236
Anti-CDH1 Rabbit Polyclonal Antibody
Antibodies
Anti-E-Cadherin Antibody

Type: Primary
Antigen: CDH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR